Assay Details
| Gene Symbol: Pwwp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127296.1 | NP_001120768.1 | 1 - 2 | 527 | 151 | |
| XM_006246135.2 | XP_006246197.1 | 1 - 2 | 1441 | 151 | ||
| XM_006246139.2 | XP_006246201.1 | 1 - 2 | 1438 | 151 | ||
| GenBank mRNA | BC166858.1 | - | 1 - 2 | 527 | 9100439 | 151 |
More Information
Updated Mapping Information
Set Membership: |
|

Pwwp2a
Genomic Map