Assay Details
| Gene Symbol: Pcgf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129882.1 | NP_001123354.1 | 2 - 3 | 406 | 66 | |
| XM_006231311.2 | XP_006231373.1 | 2 - 3 | 499 | 66 | ||
| XM_008760377.1 | XP_008758599.1 | 1 - 2 | 124 | 66 | ||
| GenBank mRNA | BC167067.1 | - | 2 - 3 | 406 | 7104558 | 66 |
| FQ214233.1 | - | 2 - 3 | 387 | 66 | ||
| FQ233837.1 | - | 2 - 3 | 340 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pcgf5
Genomic Map