Assay Details
| Gene Symbol: Zfp523 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134755.1 | NP_001128227.1 | 11 - 12 | 1437 | 105 | |
| XM_008772820.1 | XP_008771042.1 | 10 - 11 | 1420 | 105 | ||
| XM_008772821.1 | XP_008771043.1 | 10 - 11 | 1414 | 105 | ||
| GenBank mRNA | BC168249.1 | - | 11 - 12 | 1437 | 5598849 | 105 |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp523
Genomic Map