Assay Details
| Gene Symbol: Zfp523 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134755.1 | NP_001128227.1 | 9 - 10 | 1036 | 66 | |
| XM_008772820.1 | XP_008771042.1 | 8 - 9 | 1019 | 66 | ||
| XM_008772821.1 | XP_008771043.1 | 8 - 9 | 1013 | 66 | ||
| GenBank mRNA | BC168249.1 | - | 9 - 10 | 1036 | 5598849 | 66 |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp523
Genomic Map