Assay Details
| Gene Symbol: Zfp707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135903.1 | NP_001129375.1 | 4 - 5 | 1190 | 67 | |
| GenBank mRNA | BC161959.1 | - | 4 - 5 | 1190 | 9027463 | 67 |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp707
Genomic Map