Assay Details
Gene Symbol: RGD1559747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027235.1 | 1 - 2 | 156 | 66 | ||
GenBank mRNA | BE121381.1 | - | 1 - 2 | 66 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BF416827.1 | - | Chr. - - [-] on Build | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger protein 775 | Zfp775 |
RGD1304910, Znf775 |
-4.6kb | Sense | Chr. 4 - 78240325 - 78258545 on NCBI Build Rnor_6.0 |
Downstream |
GTPase, IMAP family member 8 |
Gimap8 | IanT | 18.0kb | Sense | Chr. 4 - 78283357 - 78294838 on NCBI Build Rnor_6.0 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
