Assay Details
| Gene Symbol: Pom121l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162931.1 | NP_001156403.1 | 1 - 2 | 1600 | 82 | |
| GenBank mRNA | BC097347.1 | - | 1 - 2 | 1600 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pom121l2
Genomic Map