Assay Details
Gene Symbol: Rem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022685.2 | NP_073176.2 | 3 - 4 | 607 | 79 | |
GenBank mRNA | AF084464.1 | - | 3 - 4 | 481 | 79 | |
AY916790.1 | - | 3 - 4 | 547 | 79 | ||
BC127495.1 | - | 3 - 4 | 592 | 8371030 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
