Assay Details
| Gene Symbol: Kctd16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172155.1 | NP_001165626.1 | 1 - 2 | 839 | 94 | |
| GenBank mRNA | FQ212362.1 | - | 1817 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kctd16
Genomic Map