Assay Details
Gene Symbol: Abcg3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004076.2 | NP_001004076.2 | 12 - 13 | 1535 | 83 | |
XM_006250626.2 | XP_006250688.1 | 12 - 13 | 1584 | 83 | ||
XM_008770002.1 | XP_008768224.1 | 12 - 13 | 3925 | 83 | ||
XM_008770003.1 | XP_008768225.1 | 13 - 14 | 3805 | 83 | ||
GenBank mRNA | BC098896.1 | - | 12 - 13 | 1535 | 7100556 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
