Assay Details
Gene Symbol: Abcg3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004076.2 | NP_001004076.2 | 9 - 10 | 1232 | 103 | |
XM_006250626.2 | XP_006250688.1 | 9 - 10 | 1281 | 103 | ||
XM_008770002.1 | XP_008768224.1 | 9 - 10 | 3622 | 103 | ||
XM_008770003.1 | XP_008768225.1 | 10 - 11 | 3502 | 103 | ||
GenBank mRNA | AY688113.1 | - | 8 - 9 | 1159 | 103 | |
BC098896.1 | - | 9 - 10 | 1232 | 7100556 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
