Assay Details
| Gene Symbol: Trmt13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033902.2 | NP_001029074.2 | 4 - 5 | 354 | 78 | |
| XM_008761469.1 | XP_008759691.1 | 2 - 3 | 229 | 78 | ||
| GenBank mRNA | BC103656.1 | - | 4 - 5 | 402 | 78 | |
| BC166503.1 | - | 4 - 5 | 354 | 7122476 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trmt13
Genomic Map