Assay Details
| Gene Symbol: LOC501233 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047967.1 | NP_001041432.1 | 2 - 3 | 156 | 78 | |
| GenBank mRNA | AY387066.1 | - | 2 - 3 | 156 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC501233
Genomic Map