Assay Details
| Gene Symbol: Hist1h2bo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106114.1 | NP_001099584.1 | 1 - 2 | 374 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist1h2bo| Gene Symbol: Hist1h2bo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106114.1 | NP_001099584.1 | 1 - 2 | 374 | 124 | |
Set Membership: |
|