Assay Details
| Gene Symbol: Zfp280b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106384.1 | NP_001099854.1 | 1 - 2 | 131 | 54 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106384.1 transcripts.
Set Membership: |
|

Zfp280b
Genomic Map