Assay Details
Gene Symbol: Tspo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012515.2 | NP_036647.1 | 3 - 4 | 411 | 97 | |
GenBank mRNA | BQ194961.1 | - | 1 - 2 | 97 | ||
FQ228784.1 | - | 3 - 4 | 413 | 97 | ||
J05122.1 | - | 2 - 3 | 354 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
