Assay Details
Gene Symbol: Mgp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012862.1 | NP_036994.1 | 2 - 3 | 99 | 94 | |
GenBank mRNA | BC086394.1 | - | 2 - 3 | 131 | 7303359 | 94 |
J03026.1 | - | 2 - 3 | 99 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
