Assay Details
| Gene Symbol: Rpl39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012875.2 | NP_037007.1 | 124 | 88 | ||
| GenBank mRNA | BC058489.1 | - | 158 | 6887843 | 88 | |
| X82551.1 | - | 124 | 88 | |||
| Gene Symbol: LOC103690821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773208.1 | XP_008771430.1 | 1 | 124 | 88 | |
| Gene Symbol: LOC103692785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008770106.1 | XP_008768328.1 | 1 | 158 | 88 | |
| Gene Symbol: LOC103692831 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008765320.1 | XP_008763542.1 | 1 | 154 | 88 | |
| Gene Symbol: LOC103693375 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008770613.1 | XP_008768835.1 | 1 | 157 | 88 | |
| Gene Symbol: LOC103694169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771991.1 | XP_008770213.1 | 1 | 154 | 88 | |
| Gene Symbol: LOC103694404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008772938.1 | XP_008771160.1 | 1 | 159 | 88 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rpl39
Genomic Map