Assay Details
Gene Symbol: Fabp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053365.1 | NP_445817.1 | 1 - 2 | 124 | 74 | |
GenBank mRNA | AF144756.1 | - | 1 - 2 | 124 | 74 | |
BC084721.1 | - | 1 - 2 | 112 | 7310595 | 74 | |
U75581.1 | - | 1 - 2 | 71 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
