Assay Details
| Gene Symbol: Carhsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152790.2 | NP_690003.1 | 1 - 2 | 40 | 104 | |
| XM_008767479.1 | XP_008765701.1 | 1 - 2 | 517 | 104 | ||
| GenBank mRNA | BC071173.1 | - | 1 - 2 | 40 | 7099252 | 104 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC071173.1,NM_152790.2 transcripts.
Set Membership: |
|

Carhsp1
Genomic Map