Assay Details
| Gene Symbol: Gsta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031509.2 | NP_113697.1 | 1 - 2 | 223 | 86 | |
| GenBank mRNA | AF111160.1 | - | 1 - 2 | 223 | 86 | |
| FQ213952.1 | - | 1 - 2 | 174 | 86 | ||
| S72505.1 | - | 1 - 2 | 133 | 86 | ||
| X78848.1 | - | 1 - 2 | 133 | 86 | ||
| Gene Symbol: LOC102550391 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244659.2 | XP_006244721.2 | 218 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF111160.1, NM_031509.2, S72505.1, X78848.1, XM_006244659.2 transcripts.
Set Membership: |
|

Gsta1
Genomic Map