Assay Details
| Gene Symbol: RT1-T24-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008826.3 | NP_001008826.2 | 1 | 51 | 106 | |
| GenBank mRNA | FM045430.1 | - | 1 | 51 | 106 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_001008826.3 transcripts.
This assay is in the 5'UTR region of the NM_001008826.3 transcripts.
Set Membership: |
|

RT1-T24-4
Genomic Map