Assay Details
| Gene Symbol: Mrps21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126094.2 | NP_001119566.1 | 1 | 53 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001126094.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mrps21
Genomic Map