Assay Details
| Gene Symbol: Ces1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133295.3 | NP_579829.3 | 1 | 53 | 71 | |
| GenBank mRNA | BC061789.1 | - | 1 | 53 | 5623732 | 71 |
| X51974.1 | - | 1 | 50 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC061789.1, NM_133295.3, X51974.1 transcripts.
Set Membership: |
|

Ces1d
Genomic Map