Assay Details
| Gene Symbol: Cyp4a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175837.1 | NP_787031.1 | 1 | 47 | 102 | |
| XM_006238712.2 | XP_006238774.1 | 1 | 59 | 102 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the NM_175837.1,XM_006238712.2 transcripts.
This assay is in the 5'UTR region of the NM_175837.1,XM_006238712.2 transcripts.
Set Membership: |
|

Cyp4a1
Genomic Map