Assay Details
| Gene Symbol: Ugt1a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175846.2 | NP_787040.2 | 1 | 748 | 100 | |
| GenBank mRNA | AF461735.1 | - | 1 | 751 | 100 | |
| AY435135.1 | - | 1 | 748 | 100 | ||
| Gene Symbol: LOC102553187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008767335.1 | XP_008765557.1 | 2 | 3587 | 100 | |
| XM_008767336.1 | XP_008765558.1 | 2 | 3486 | 100 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ugt1a8
Genomic Map