Assay Details
| Gene Symbol: Pcdha7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199507.1 | NP_955801.1 | 1 | 1145 | 114 | |
| GenBank mRNA | AB113390.1 | - | 1 | 1145 | 114 | |
| AY573981.1 | - | 1 | 1145 | 114 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pcdha7
Genomic Map