Assay Details
| Gene Symbol: Mir103-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031860.1 | 1 | 45 | 78 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitochondrial antiviral signaling protein | Mavs | Visa | -43.1kb | Sense | Chr. 3 - 123776154 - 123790572 on NCBI Build Rnor_6.0 |
| Downstream | ring finger protein 24 | Rnf24 | 15.1kb | Anti-sense | Chr. 3 - 123848825 - 123849788 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir103-2
Genomic Map