Assay Details
| Gene Symbol: Mir877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032258.1 | 1 | 40 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ATP-binding cassette, subfamily F (GCN20), member 1 |
Abcf1 | Abc50 | Sense | Chr. 20 - 3309947 - 3322828 on NCBI Build Rnor_6.0 | |
| Downstream |
protein phosphatase 1, regulatory subunit 10 |
Ppp1r10 |
Fb19, Pnuts |
10.7kb | Anti-sense | Chr. 20 - 3329677 - 3344768 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir877
Genomic Map