Assay Details
| Gene Symbol: Mir30d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031842.1 | 1 | 43 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-30b | Mir30b | rno-mir-30b | -3.7kb | Sense | Chr. 7 - 109282662 - 109282747 on NCBI Build Rnor_6.0 |
| Downstream |
similar to H3 histone, family 3B |
RGD1564548 | 124.9kb | Anti-sense | Chr. 7 - 109411475 - 109411885 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir30d
Genomic Map