Assay Details
| Gene Symbol: Mir146a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031892.1 | 1 | 51 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ATPase, class V, type 10B |
Atp10b | RGD1565126 | -151.6kb | Anti-sense | Chr. 10 - 28684914 - 28810835 on NCBI Build Rnor_6.0 |
| Downstream | pituitary tumor-transforming 1 | Pttg1 | Pttg | 51.8kb | Sense | Chr. 10 - 29014367 - 29026088 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir146a
Genomic Map