Assay Details
Gene Symbol: Mir187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031905.1 | 1 | 54 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Ab2-143 | LOC498829 | -196.6kb | Sense | Chr. 18 - 16189822 - 16193861 on NCBI Build Rnor_6.0 | |
Downstream | protein NYNRIN-like | LOC102553405 | 21.8kb | Anti-sense | Chr. 18 - 16412420 - 16413493 on NCBI Build Rnor_6.0 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
