Assay Details
| Gene Symbol: Mir211 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031924.1 | 1 | 56 | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Kruppel-like factor 13 | Klf13 | RGD1565099 | -238.8kb | Anti-sense | Chr. 1 - 124772596 - 124803363 on NCBI Build Rnor_6.0 |
| Downstream | myotubularin related protein 10 | Mtmr10 | RGD1305958 | 82.5kb | Sense | Chr. 1 - 125124743 - 125175857 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir211
Genomic Map