Assay Details
| Gene Symbol: Mir377 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032114.1 | 1 | 29 | 63 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 496 | Mir496 | rno-mir-496 | -1.3kb | Sense | Chr. 6 - 133889283 - 133889361 on NCBI Build Rnor_6.0 |
| Downstream | microRNA 541 | Mir541 | rno-mir-541 | 1.9kb | Sense | Chr. 6 - 133892655 - 133892744 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir377
Genomic Map