Assay Details
| Gene Symbol: Cb707485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029398.1 | 2 - 3 | 219 | 72 | ||
| GenBank mRNA | CB707485.1 | - | 2 - 3 | 232 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | immediate early response 3 | Ier3 |
Dif2, Iex1, Ler3, Prg1 |
-92.2kb | Sense | Chr. 20 - 3438798 - 3440002 on NCBI Build Rnor_6.0 |
| Downstream | discoidin domain receptor tyrosine kinase 1 | Ddr1 |
Cak, Drd1, PTK3D |
22.4kb | Anti-sense | Chr. 20 - 3555135 - 3574959 on NCBI Build Rnor_6.0 |
More Information
Updated Mapping Information
Set Membership: |
|

Cb707485
Genomic Map