Assay Details
Gene Symbol: Mir7a-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031795.1 | 1 | 48 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RecQ mediated genome instability 1 | Rmi1 | RGD1310671 | -9.4kb | Anti-sense | Chr. 17 - 6658285 - 6665619 on NCBI Build Rnor_6.0 |
Downstream | similar to CG9752-PA | RGD1311345 | 8.0kb | Sense | Chr. 17 - 6683152 - 6690724 on NCBI Build Rnor_6.0 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
