Assay Details
| Gene Symbol: Mir135b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031797.1 | 1 | 46 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cyclin-dependent kinase 18 | Cdk18 |
PCTAIRE3, Pctk3 |
-50.3kb | Anti-sense | Chr. 13 - 48893522 - 48926689 on NCBI Build Rnor_6.0 |
| Downstream | LEM domain-containing protein 1-like | LOC102556277 | 26.7kb | Sense | Chr. 13 - 49003763 - 49035519 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir135b
Genomic Map