Assay Details
Gene Symbol: Mir20b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032137.1 | 1 | 37 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 19b-2 | Mir19b2 |
Mir19b-2, rno-mir-19b-2 |
-44.0 | Sense | Chr. X - 140117379 - 140117474 on NCBI Build Rnor_6.0 |
Downstream | microRNA 17-2 | Mir17-2 | rno-mir-17-2 | 302.0 | Sense | Chr. X - 140117891 - 140117968 on NCBI Build Rnor_6.0 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
