Assay Details
| Gene Symbol: Mir708 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032291.1 | 1 | 40 | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
family with sequence similarity 181, member B |
Fam181b | RGD1564695 | -3.3mb | Sense | Chr. 1 - 157920786 - 157922283 on NCBI Build Rnor_6.0 |
| Downstream | teneurin transmembrane protein 4 | Tenm4 | Odz4 | 106.1kb | Sense | Chr. 1 - 161327440 - 161885094 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir708
Genomic Map