Assay Details
| Gene Symbol: Mir568 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032745.1 | 1 | 52 | 93 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | T cell immunoreceptor with Ig and ITIM domains | Tigit | RGD1563191 | -8.3kb | Anti-sense | Chr. 11 - 61970617 - 61987749 on NCBI Build Rnor_6.0 |
| Downstream | zinc finger and BTB domain containing 20 | Zbtb20 | RGD1560387 | 18.8kb | Sense | Chr. 11 - 62014925 - 62561113 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir568
Genomic Map