Assay Details
| Gene Symbol: Mir653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_032751.1 | 1 | 46 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 132 | Ccdc132 | RGD1304751 | -107.1kb | Anti-sense | Chr. 4 - 28441264 - 28551784 on NCBI Build Rnor_6.0 |
| Downstream | microRNA mir-489 | Mir489 | rno-mir-489 | 545.0 | Sense | Chr. 4 - 28659523 - 28659628 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir653
Genomic Map