Assay Details
| Gene Symbol: Mir3547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037321.1 | 1 | 55 | 54 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
TSR3, 20S rRNA accumulation |
Tsr3 | RGD1565744 | -2.6kb | Anti-sense | Chr. 10 - 14598014 - 14600576 on NCBI Build Rnor_6.0 |
| Downstream | ubiquitin-conjugating enzyme E2I | Ube2i | UbcE2A | 15.4kb | Sense | Chr. 10 - 14618612 - 14639285 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir3547
Genomic Map