Assay Details
| Gene Symbol: Mir3597-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037359.1 | 1 | 61 | 100 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-9-2 | Mir9-2 |
Mir9a-2, rno-mir-9-2, rno-mir-9a-2 |
Anti-sense | Chr. 2 - 11884671 - 11884757 on NCBI Build Rnor_6.0 | |
| Downstream |
centrin, EF-hand protein, 3 |
Cetn3 | 216.7kb | Anti-sense | Chr. 2 - 12101486 - 12115009 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir3597-2
Genomic Map