Assay Details
| Gene Symbol: Mir2985 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037389.1 | 1 | 52 | 74 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
family with sequence similarity 198, member B |
Fam198b | Ened | -841.3kb | Anti-sense | Chr. 2 - 178678940 - 178750054 on NCBI Build Rnor_6.0 |
| Downstream |
glycine receptor, beta |
Glrb | 176.6kb | Sense | Chr. 2 - 179768040 - 179842612 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir2985
Genomic Map