Assay Details
| Gene Symbol: Mir98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031855.1 | 1 | 58 | 90 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA let-7f-2 | Mirlet7f-2 | rno-let-7f-2 | -521.0 | Sense | Chr. X - 21583272 - 21583354 on NCBI Build Rnor_6.0 |
| Downstream | hydroxysteroid (17-beta) dehydrogenase 10 | Hsd17b10 | Hadh2 | 112.8kb | Sense | Chr. X - 21696796 - 21699241 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir98
Genomic Map