Assay Details
| Gene Symbol: Mir99a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031856.1 | 1 | 43 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin specific peptidase 25 | Usp25 | -493.5kb | Sense | Chr. 11 - 15436048 - 15558646 on NCBI Build Rnor_6.0 | |
| Downstream | microRNA let-7c-1 | Mirlet7c-1 | rno-let-7c-1 | 640.0 | Sense | Chr. 11 - 16052873 - 16052966 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir99a
Genomic Map