Assay Details
| Gene Symbol: Mirlet7c-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031803.1 | 1 | 49 | 90 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 99a | Mir99a | rno-mir-99a | -640.0 | Sense | Chr. 11 - 16052153 - 16052233 on NCBI Build Rnor_6.0 |
| Downstream | microRNA 3588 | Mir3588 | rno-mir-3588 | 44.4kb | Anti-sense | Chr. 11 - 16097337 - 16097442 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mirlet7c-1
Genomic Map