Assay Details
| Gene Symbol: Mir152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031894.1 | 1 | 45 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
coatomer protein complex, subunit zeta 2 |
Copz2 | Sense | Chr. 10 - 84718824 - 84731880 on NCBI Build Rnor_6.0 | ||
| Downstream | CDK5 regulatory subunit associated protein 3 | Cdk5rap3 | C53 | 61.2kb | Anti-sense | Chr. 10 - 84780646 - 84789787 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir152
Genomic Map