Assay Details
| Gene Symbol: Mir3560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037342.1 | 1 | 58 | 103 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-362 | Mir362 | rno-mir-362 | -629.0 | Anti-sense | Chr. X - 16117170 - 16117234 on NCBI Build Rnor_6.0 |
| Downstream | microRNA mir-500 | Mir500 | rno-mir-500 | 3.4kb | Anti-sense | Chr. X - 16121332 - 16121411 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir3560
Genomic Map