Assay Details
| Gene Symbol: Mir3567 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037353.1 | 1 | 65 | 80 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BI395608.1 | - | Chr. - - [-] on Build | 80 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-126 | Mir126 |
Mir126a, rno-mir-126, rno-mir-126a |
Anti-sense | Chr. 3 - 4042488 - 4042560 on NCBI Build Rnor_6.0 | |
| Downstream | 1-acylglycerol-3-phosphate O-acyltransferase 2 | Agpat2 | 2.2kb | Sense | Chr. 3 - 4044741 - 4055384 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir3567
Genomic Map